Order Entry
Canada
Orders LinkContactUsLinkComponent
Difco™ Select APS™ Super Broth Base, BD Diagnostics

Difco™ Select APS™ Super Broth Base, BD Diagnostics

Difco™ Select APS™ Super Broth Base, 10 kg
Catalog # CA90000-678
Supplier:   BD
Supplier #:  212486
Difco™ Select APS™ Super Broth Base, BD Diagnostics
Catalog # CA90000-678
Supplier:  BD
CAS Number:  
Difco™ Select APS™ Super Broth Base, BD Diagnostics

Difco™ Select APS™ Super Broth Base, 10 kg

CAS Number:  
Price
$2,810.00
Log in for availability
Each (10kg)
Product Details & Documents

Select APS™ Super Broth is a molecular genetics medium used to grow Escherichia coli to a high cell density.

  • Molecular genetics medium
  • Alternative to classical animal-based media
  • Allows maintenance and propogation of E. coli
  • Soy hydrolysate provides nitrogen and carbon
  • Glycerol is added as a carbon and energy source

Select APS™ media are manufactured from animal-free ingredients in order to minimize the risk of bovine spongiform encephalopathy in culture media containing animal, and especially bovine, materials.

Specifications

Specifications

  • Description:
    Difco™ Select APS™ Super Broth Base
  • Size:
    10 kg

Specifications