Order Entry
Canada
Orders LinkContactUsLinkComponent
MacConkey Agar, BD

MacConkey Agar, BD

MacConkey Agar, 25 lbs.
  CA90000-172
 :   BD
Supplier #:  211391
MacConkey Agar, BD
  CA90000-172
 :  BD
 :  
MacConkey Agar, BD
MacConkey Agar, BD

MacConkey Agar, 25 lbs.

 :  
 
$4,030.00
   
Each (25lb)
 

MacConkey agar is a selective and differential culture medium for bacteria designed to selectively isolate Gram-negative and enteric bacilli and differentiate them based on lactose fermentation.

  • Isolates Gram-negative bacilli

Enteric bacteria that have the ability to ferment lactose can be detected using the carbohydrate lactose, and the pH indicator neutral red.

 

 

  • Description:
    MacConkey Agar
  • Size:
    25 lbs.

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.