Order Entry
Canada
Orders LinkContactUsLinkComponent
Uridine

Uridine

25 g
Catalog # CA89130-788
Supplier:   HiMedia
Supplier #:  TC181-25G
Uridine
Catalog # CA89130-788
Supplier:  HiMedia
CAS Number:  58-96-8
Uridine

25 g

CAS Number:  58-96-8
Price
$1,411.83
Log in for availability
Each (25g)
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Synonyms:  1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 1-β-D-Ribofuranosyluracil, Uracil-1-β-D-ribofuranoside
  • CAS Number:58-96-8
  • Formula:C₉H₁₂N₂O₆
  • MDL Number:MFCD00006526
  • Molecular Weight:244.20 g/mol
Specifications

Specification Test Results

  • Molecular Weight:
    244.2

Specifications

  • Size:
    25 g

Specifications