Orders LinkContactUsLinkComponent
Slimsette™ Tissue Processing/Embedding Cassettes, Simport
Aqua
  CA82028-764
 :  M509-12
Slimsette™ Tissue Processing/Embedding Cassettes, Simport
  CA82028-764
Slimsette™ Tissue Processing/Embedding Cassettes, Simport
Slimsette™ Tissue Processing/Embedding Cassettes, Simport
Aqua
$493.64
Case of 1,500
These compact, easy-to-use cassettes incorporate a unique recessed cover which allows more cassettes to be stacked
They are molded from acetal polymer to keep specimens safely submerged in liquid and are resistant to chemical actions of histological solvents. The 1x 5mm slotted openings ensure efficient fluid exchange and drainage, and the one-piece integral lid can be opened and closed as often as necessary. The anterior writing area is slanted at a 45° angle
 : CE marked.
 : Packaged in three dispenser boxes of 500 cassettes with covers assembled.
  • Color:
    Aqua
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.