Slimsette™ Tissue Processing/Embedding Cassettes, Simport
Product Details & Documents
These compact, easy-to-use cassettes incorporate a unique recessed cover which allows more cassettes to be stacked
They are molded from acetal polymer to keep specimens safely submerged in liquid and are resistant to chemical actions of histological solvents. The 1x 5mm slotted openings ensure efficient fluid exchange and drainage, and the one-piece integral lid can be opened and closed as often as necessary. The anterior writing area is slanted at a 45° angle
Certifications: CE marked.
Ordering information: Packaged in three dispenser boxes of 500 cassettes with covers assembled.
Documents
Certificates
Search by Lot NumberSpecifications
- Color:Orange
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)