Order Entry
Canada
Orders LinkContactUsLinkComponent
 

G-Biosciences Hybridization Buffer (Hybridization Denaturing Solution)

Hybridization denaturing solution (1.5 M NaCl, 0.5 N NaOH), 1 L
Catalog # CA82021-478
Supplier:   G-Biosciences
 :  R016
G-Biosciences Hybridization Buffer (Hybridization Denaturing Solution)
Catalog # CA82021-478
Supplier:  G-Biosciences
CAS Number:  
G-Biosciences Hybridization Buffer (Hybridization Denaturing Solution)
G-Biosciences Hybridization Buffer (Hybridization Denaturing Solution)

Hybridization denaturing solution (1.5 M NaCl, 0.5 N NaOH), 1 L

CAS Number:  
Price
$459.06
Log in for availability
Each (4l)
 
Product Details & Documents

Ready-to-Use: Convenient, time-saving formulation ideal for consistent experimental reproducibility.

  • 1.5 M sodium chloride

  • 0.5 M  sodium hydroxide

A high-pH reagent used to denature double-stranded nucleic acids into single strands, preparing DNA or RNA for hybridization. Ready-to-use: Convenient, time-saving formulation ideal for consistent experimental reproducibility.

Specifications

Specifications

  • Description:
    Hybridization denaturing solution (1.5 M NaCl, 0.5 N NaOH)
  • Size:
    1 L

Specifications