Order Entry
Canada
Orders LinkContactUsLinkComponent
G-Biosciences Hybridization Buffer (Hybridization Denaturing Solution)

G-Biosciences Hybridization Buffer (Hybridization Denaturing Solution)

Hybridization denaturing solution (1.5 M NaCl, 0.5 N NaOH), 1 gallon
Catalog # CA82021-476
Supplier:   G-Biosciences
Supplier #:  R015
G-Biosciences Hybridization Buffer (Hybridization Denaturing Solution)
Catalog # CA82021-476
Supplier:  G-Biosciences
CAS Number:  
G-Biosciences Hybridization Buffer (Hybridization Denaturing Solution)
G-Biosciences Hybridization Buffer (Hybridization Denaturing Solution)

Hybridization denaturing solution (1.5 M NaCl, 0.5 N NaOH), 1 gallon

CAS Number:  
Price
$292.63
Log in for availability
Each (1l)
Minimum order quantity is 1
Product Details & Documents

Ready-to-Use: Convenient, time-saving formulation ideal for consistent experimental reproducibility.

  • 1.5 M sodium chloride

  • 0.5 M  sodium hydroxide

A high-pH reagent used to denature double-stranded nucleic acids into single strands, preparing DNA or RNA for hybridization. Ready-to-use: Convenient, time-saving formulation ideal for consistent experimental reproducibility.

Specifications

Specifications

  • Description:
    Hybridization denaturing solution (1.5 M NaCl, 0.5 N NaOH)
  • Size:
    1 gallon

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.