Order Entry
Canada
Orders LinkContactUsLinkComponent
Polyester Printer Labels, Brady

Polyester Printer Labels, Brady

Tls print ribbon blue, Blue, TLS 2200®, TLS PC Link®, BMP®71, 2", 75 ft.
  CA80090-204
Polyester Printer Labels, Brady
  CA80090-204
 :  
Polyester Printer Labels, Brady
Polyester Printer Labels, Brady

Tls print ribbon blue, Blue, TLS 2200®, TLS PC Link®, BMP®71, 2", 75 ft.

 
 

B-461: Self-laminating polyester is a clear film with a matte white printable zone ideal for cables, laboratory ID or general labeling where graphics need to be protected.
B-490: Durable polyester markers are designed for use in laboratory identification applications such as vials, centrifuge, tubes, test tubes, straws and slides.

B-488: High performance polyester withstands numerous solvents and variable temperatures when applied to various surfaces including stainless steel and polypropylene.
B-498: Vinyl cloth backing offers excellent holding power and offers clean removal and repositioning.

B-424: Paper labels with permanent rubber adhesive provide good contrast and smear resistance for thermal transfer and barcode printing applications.
B-427: Self-laminating vinyl is an excellent material for wire and cable identification. This material has good clarity, conformability and is self-extinguishing.

B-430: Clear polyester labels are designed for applications that use barcodes, alphanumeric, graphic symbols and logos and require nameplate-like quality.
B-499: Nylon cloth wrap-around labels are highly pliable to conform to tightly curved surfaces such as wire and cable.

B-351: Tamper resistant vinyl is designed to fracture easily to show signs of product tampering and to prevent on-piece label removal.
B-423: Permanent polyester is ideal for electronic PCB and component identification, bar code labels, rating plates and solar panel identification.
B-439: Permanent vinyl is a thermal transfer printable material ideal for applications needing various colors like finished product, rating plates and general-purpose printers.

Certifications: RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC.

 
  • For Use With:
    TLS 2200®, TLS PC Link®, BMP®71
  • Color:
    Blue
  • Description:
    Tls print ribbon blue
  • Roll length:
    75 ft.
  • Width:
    2"
Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.