Orders LinkContactUsLinkComponent
Labels, Brady
Printer labels, FNAHSL properties, Polyester, 0.2 ml tubes, 38.1 x 4.8 x 17.8 mm
  CA80090-134
 :   Avantor
Labels, Brady
  CA80090-134
 :  Avantor
Labels, Brady
Labels, Brady
Printer labels, FNAHSL properties, Polyester, 0.2 ml tubes, 38.1 x 4.8 x 17.8 mm
$117.85
Each
B-499: Nylon Cloth wrap-around labels are highly pliable to conform to tightly curved surfaces such as wire and cable. This material has a permanent acrylic adhesive and is thermal transfer printable.
B-488: High Performance Polyester withstands numerous solvents and variable temperatures when applied to various surfaces including Stainless Steel and Polypropylene.
B-461: Self-Laminating Polyester is a clear film with a matte white printable zone ideal for cables, laboratory ID or general labeling where graphics need to be protected.
B-427: Self-Laminating Vinyl is an excellent material for wire and cable identification. This material has good clarity, conformability and is self-extinguishing.
B-351: Tamper-Resistant Vinyl is designed to fracture easily in order to show signs of product tampering and to prevent on-piece label removal.
 : RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC; UL approved.
  • For Use With:
    0.2 ml tubes
  • Description:
    Printer labels, FNAHSL properties
  • Dimensions:
    38.1 x 4.8 x 17.8 mm
  • Material:
    Polyester
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.