Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Polyester Printer Labels, Brady

Tls printer labels B-490 MAT´L, Polyester, White, BMP®61 labeling system, BMP®71 label printer, TLS 2200® thermal labeling system, TLS-PC LINK™ thermal labeling system, BradyPrinter™ M611, 1.5×3.875"
Catalog # CA80090-118
Supplier:   Avantor
Polyester Printer Labels, Brady
Catalog # CA80090-118
Supplier:  Avantor
Polyester Printer Labels, Brady
Polyester Printer Labels, Brady

Tls printer labels B-490 MAT´L, Polyester, White, BMP®61 labeling system, BMP®71 label printer, TLS 2200® thermal labeling system, TLS-PC LINK™ thermal labeling system, BradyPrinter™ M611, 1.5×3.875"

Price
$97.84
Log in for availability
Each
Product Details & Documents

B-461: Self-laminating polyester is a clear film with a matte white printable zone ideal for cables, laboratory ID or general labeling where graphics need to be protected.
B-490: Durable polyester markers are designed for use in laboratory identification applications such as vials, centrifuge, tubes, test tubes, straws and slides.

B-488: High performance polyester withstands numerous solvents and variable temperatures when applied to various surfaces including stainless steel and polypropylene.
B-498: Vinyl cloth backing offers excellent holding power and offers clean removal and repositioning.

B-424: Paper labels with permanent rubber adhesive provide good contrast and smear resistance for thermal transfer and barcode printing applications.
B-427: Self-laminating vinyl is an excellent material for wire and cable identification. This material has good clarity, conformability and is self-extinguishing.

B-430: Clear polyester labels are designed for applications that use barcodes, alphanumeric, graphic symbols and logos and require nameplate-like quality.
B-499: Nylon cloth wrap-around labels are highly pliable to conform to tightly curved surfaces such as wire and cable.

B-351: Tamper resistant vinyl is designed to fracture easily to show signs of product tampering and to prevent on-piece label removal.
B-423: Permanent polyester is ideal for electronic PCB and component identification, bar code labels, rating plates and solar panel identification.
B-439: Permanent vinyl is a thermal transfer printable material ideal for applications needing various colors like finished product, rating plates and general-purpose printers.

 : RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC.

Specifications
  • For Use With:
    BMP®61 labeling system, BMP®71 label printer, TLS 2200® thermal labeling system, TLS-PC LINK™ thermal labeling system, BradyPrinter™ M611
  • Color:
    White
  • Description:
    Tls printer labels B-490 MAT´L
  • Dimensions:
    1.5×3.875"
  • Material:
    Polyester