Orders LinkContactUsLinkComponent
Labels, Brady
Labels, Brady
Portable thermal labels, Nylon Cloth, 12.7×38.1 mm (0.5×1.5"), 500 labels/roll
Catalog # CA80090-086
Supplier:   Avantor
Labels, Brady
Catalog # CA80090-086
Supplier:  Avantor
Labels, Brady
Labels, Brady
Portable thermal labels, Nylon Cloth, 12.7×38.1 mm (0.5×1.5"), 500 labels/roll
Price
$103.40
Log in for availability
Each
Product Details & Documents
Labels for the TLS 2200 and TLS-PC Link Thermal Transfer Portable Printers.
B-488: High Performance Polyester withstands numerous solvents and variable temperatures when applied to various surfaces including Stainless Steel and Polypropylene.
B-461: Self-Laminating Polyester is a clear film with a matte white printable zone ideal for cables, laboratory ID or general labeling where graphics need to be protected.
B-499: Nylon Cloth wrap-around labels are highly pliable to conform to tightly curved surfaces such as wire and cable.
Please Note: This material is UL, CSA and AGA approved with its respective ribbon. See the 'Recommended Ribbon Series' for the appropriate ribbon to use.
Specifications
  • Description:
    Portable thermal labels
  • Dimensions:
    12.7×38.1 mm (0.5×1.5")
  • Material:
    Nylon Cloth
  • Packaging:
    500 labels/roll
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.