Order Entry
Canada
Orders LinkContactUsLinkComponent
Propidium Iodide, Calbiochem®

Propidium Iodide, Calbiochem®

Glass bottle, 50 mg
Catalog # CA80057-370
Irritant
Supplier:   MilliporeSigma
Supplier #:  537059-50MG
Propidium Iodide, Calbiochem®
Catalog # CA80057-370
Supplier:  MilliporeSigma
CAS Number:  25535-16-4
Propidium Iodide, Calbiochem®

Glass bottle, 50 mg

CAS Number:  25535-16-4
Price
$385.16
Log in for availability
Each (50mg)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Membrane-impermeable DNA intercalator. Has red fluorescence at 488nm. Useful for flow cytometry for staining apoptotic cells and nuclei. Can be used to differentiate between apoptotic and necrotic cell death. Can be used together with common green fluorescent probes labeled with fluorescein isothiocyanate.

Extinction coefficient (493nm): 5900M-1cm-1; (287nm): 55,700M-1cm-1. Excitation maximum: 536nm. Emission maximum: 617nm.

Synonyms:  3,8-Diamino-5-[3-(diethylmethylammonio)propyl]-6-phenylphenanthridinium diiodide, PI, 3,8-Diamino-5-(3-(diethyl(methyl)ammonio)propyl)-6-phenylphenanthridin-5-ium iodide, 3,8-Diamino-5-(3-diethylaminopropyl)-6-phenylphenanthridinium iodide methiodide
  • CAS Number:25535-16-4
  • Formula:C₂₇H₃₄I₂N₄
  • MDL Number:MFCD00011921
  • Molecular Weight:668.4 g/mol
  • Storage Temperature:Refrigerator
  • EINECS:247-081-0

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Size:
    50 mg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.