Order Entry
Canada
Orders LinkContactUsLinkComponent
Sodium hydroxide, pellets

Sodium hydroxide, pellets

Metal can, 250 g
Catalog # CA80055-562
Supplier:   MilliporeSigma
Supplier #:  567530-250GM
Sodium hydroxide, pellets
Catalog # CA80055-562
Supplier:  MilliporeSigma
CAS Number:  1310-73-2
Sodium hydroxide, pellets

Metal can, 250 g

CAS Number:  1310-73-2
Price
$247.91
Log in for availability
Each (250g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol

Documents

Certificates
Search by Lot Number
Specifications

Specifications

  • Pack type:
    Metal can
  • Size:
    250 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.