Orders LinkContactUsLinkComponent
Histosette® I Tissue Processing/Embedding Cassettes, Simport
Histosette® I Tissue Processing/Embedding Cassettes, Simport
Fluorescent Pink
Catalog # CA73960-524
Supplier:   Simport Scientific
Supplier #:  M498-16
Histosette® I Tissue Processing/Embedding Cassettes, Simport
Catalog # CA73960-524
Supplier:  Simport Scientific
Histosette® I Tissue Processing/Embedding Cassettes, Simport
Histosette® I Tissue Processing/Embedding Cassettes, Simport
Fluorescent Pink
Price
$370.57
Log in for availability
Case of 1,500
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These disposable cassettes hold tissue specimens efficiently during the embedding process and in storage
Molded from acetal polymer, they keep specimens submerged in liquid and are resistant to the chemical action of histological solvents. The efficient flow-through slots maximize fluid exchange and ensure proper drainage. The one-piece integral lids snap apart and lock easily into the base of the cassettes. The anterior writing area is angled at 45° for suitability with certain types of cassette labeling instruments.
Ordering information: Packed in three dispenser boxes of 500 cassettes. Fluorescent colored cassettes require a minimum order quantity of 40 cases for purchase.
Specifications
  • Color:
    Fluorescent Pink
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)