Orders LinkContactUsLinkComponent
Capinsert™ Color Coding Inserts for Cryovial® Tubes, Simport Scientific
Capinsert™ Color Coding Inserts for Cryovial® Tubes, Simport Scientific
Green
Catalog # CA73960-408
Supplier:   Simport Scientific
Supplier #:  T312-4
Capinsert™ Color Coding Inserts for Cryovial® Tubes, Simport Scientific
Catalog # CA73960-408
Supplier:  Simport Scientific
Capinsert™ Color Coding Inserts for Cryovial® Tubes, Simport Scientific
Capinsert™ Color Coding Inserts for Cryovial® Tubes, Simport Scientific
Green
Price
$30.73
Log in for availability
Pack of 500
Product Details & Documents
Color coded inserts fit precisely into the cap of the Cryovial® tubes for color identification.
Made of polypropylene
Ordering information: T312-7 contains 100 pieces each of white, blue, red, green and yellow inserts packaged in individual bags.
Specifications
  • Color:
    Green