Orders LinkContactUsLinkComponent
Polypropylene Vials, Starplex®
Polypropylene Vials, Starplex®
Labeled, 10.00 ml, Flat bottom, Clear, Yes, Screw cap, attached, in plastic peel pack, White, Sterile
Catalog # CA73270-066
Supplier:   Starplex
Supplier #:  V102-POUCH
Polypropylene Vials, Starplex®
Catalog # CA73270-066
Supplier:  Starplex
Polypropylene Vials, Starplex®
Polypropylene Vials, Starplex®
Labeled, 10.00 ml, Flat bottom, Clear, Yes, Screw cap, attached, in plastic peel pack, White, Sterile
Price
$350.00
Log in for availability
Case of 100
Product Details & Documents
These vials are made of crack-resistant medical grade polypropylene.
  • With or without screw caps
  • Flat, conical or round bottoms
Available clear for easy visual inspection and amber for light-sensitive components. These vials provide convenient storage for samples.
Specifications
  • Cap included:
    Yes
  • Cap type:
    Screw cap, attached, in plastic peel pack
  • Sterility:
    Sterile
  • Capacity:
    10.00 ml
  • Color:
    Clear
  • Description:
    Labeled
  • Cap color:
    White
  • Bottom style:
    Flat bottom