Orders LinkContactUsLinkComponent
Polypropylene Vials, Starplex®
Unlabeled, 10.00 ml, Flat bottom, Amber, Yes, Screw cap, packed separately, White, Non sterile
Catalog # CA73240-133
Supplier:   Starplex
 :  V103-NL
Polypropylene Vials, Starplex®
Catalog # CA73240-133
Supplier:  Starplex
Polypropylene Vials, Starplex®
Polypropylene Vials, Starplex®
Unlabeled, 10.00 ml, Flat bottom, Amber, Yes, Screw cap, packed separately, White, Non sterile
Price
$397.00
Log in for availability
Case of 1,000
Product Details & Documents
These vials are made of crack-resistant medical grade polypropylene.
  • With or without screw caps
  • Flat, conical or round bottoms
Available clear for easy visual inspection and amber for light-sensitive components. These vials provide convenient storage for samples.
Specifications
  • Cap included:
    Yes
  • Cap type:
    Screw cap, packed separately
  • Sterility:
    Non sterile
  • Capacity:
    10.00 ml
  • Color:
    Amber
  • Description:
    Unlabeled
  • Cap color:
    White
  • Bottom style:
    Flat bottom