Polypropylene Vials, Starplex®
Labeled, 10.00 ml, Flat bottom, Clear, Yes, Screw cap, packed separately, White, Non sterile
Catalog # CA73240-129
Supplier: Starplex
: V102
Jump to:
Product Details & Documents
These vials are made of crack-resistant medical grade polypropylene.
- With or without screw caps
- Flat, conical or round bottoms
Available clear for easy visual inspection and amber for light-sensitive components. These vials provide convenient storage for samples.
Specifications
- Cap included:Yes
- Cap type:Screw cap, packed separately
- Sterility:Non sterile
- Capacity:10.00 ml
- Color:Clear
- Description:Labeled
- Cap color:White
- Bottom style:Flat bottom



