Order Entry
Canada
Orders LinkContactUsLinkComponent
CrossLab Split Liners for GC, Agilent Technologies | Avantor

CrossLab Split Liners for GC, Agilent Technologies

Inlet liner, split, straight, with cup (for manual injections), wool, 6.25 mm, 800 µl, 4×78.5 mm
Catalog # CA5183-4699
Fournisseur #:  5183-4699
CrossLab Split Liners for GC, Agilent Technologies
Catalog # CA5183-4699
CrossLab Split Liners for GC, Agilent Technologies
CrossLab Split Liners for GC, Agilent Technologies

Inlet liner, split, straight, with cup (for manual injections), wool, 6.25 mm, 800 µl, 4×78.5 mm

Price
$350.00
Log in for availability
Pack of 5
Product Details & Documents

Agilent split liners are made to strict dimensional specifications for optimal inlet performance.

  • Feature Agilent’s exclusive deactivation;Agilent recommends part number 5183-4647 as the top split liner choice

Split injections are intended for use with a split/splitless injection port in gas chromatography analysis. Split liners tend to have a slightly smaller outer diameter to allow for gas flow. For ease of use and reproducibility, the liners have a positioning bead, a restriction to secure the position of the glass wool, and a feature to consistently self-position to the recommended height.;Agilent split liners are made to strict dimension specifications for optimal inlet performance and feature the tightest tolerances for od, id, taper and glass wool placement.

Specifications
  • Ext.Ø:
    6.25 mm
  • Volume:
    800 µl
  • Description:
    Inlet liner, split, straight, with cup (for manual injections), wool
  • I.D.×L:
    4×78.5 mm
Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.