Histosette® I Tissue Processing/Embedding Cassettes, Simport
These disposable cassettes hold tissue specimens efficiently during the embedding process and in storage
Molded from acetal polymer, they keep specimens submerged in liquid and are resistant to the chemical action of histological solvents. The efficient flow-through slots maximize fluid exchange and ensure proper drainage. The one-piece integral lids snap apart and lock easily into the base of the cassettes. The anterior writing area is angled at 45° for suitability with certain types of cassette labeling instruments.
: Packed in three dispenser boxes of 500 cassettes. Fluorescent colored cassettes require a minimum order quantity of 40 cases for purchase.
- Color:Tan
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.