Order Entry
Canada
Orders LinkContactUsLinkComponent
Histosette® I Processing and Embedding Cassettes, Simport

Histosette® I Processing and Embedding Cassettes, Simport

Tissue Cassettes, Aqua
Catalog # CA48648-100
Supplier:   Simport Scientific
Supplier #:  M490-12
Histosette® I Processing and Embedding Cassettes, Simport
Catalog # CA48648-100
Supplier:  Simport Scientific
Histosette® I Processing and Embedding Cassettes, Simport
Histosette® I Processing and Embedding Cassettes, Simport

Tissue Cassettes, Aqua

Price
$523.39
Log in for availability
Case of 1,500
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

These disposable plastic cassettes with lids are made of acetal polymer and hold tissue or biopsy specimens during the embedding process and for storage.

They keep specimens safely submerged in liquid and are totally resistant to the chemical action of histological solvents. The efficient flow-through slots maximize fluid exchange and ensure proper drainage. The one-piece integral lid eliminates the need for separate steel lids. The lid can be easily snapped apart and locked into the base of the cassette. Lids can be opened and closed as often as necessary and always relock securely without danger of specimen loss. Large labeling areas are located on three sides of the cassette. The anterior writing area is at a 30° angle. The biopsy cassettes feature 1mm square openings.

Ordering information: Cassettes are available in 11 non cytotoxic, non metallic colors, and three fluorescent colors. Each case contains three dispenser boxes of 500 cassettes.

Documents

Specifications
  • Color:
    Aqua
  • Description:
    Tissue Cassettes

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.