Orders LinkContactUsLinkComponent
Corning® Minimum Essential Medium (Mod.) (MEM), Corning
Corning® Minimum Essential Medium (Mod.) (MEM), Corning
Minimum Essential Medium (MEM), powder with Earle's salts, without L-glutamine, phenol red, and sodium bicarbonate
Catalog # CA45000-586
Supplier:   Corning
Supplier #:  90-009-PB
Corning® Minimum Essential Medium (Mod.) (MEM), Corning
Catalog # CA45000-586
Supplier:  Corning
CAS Number:  
Corning® Minimum Essential Medium (Mod.) (MEM), Corning
Corning® Minimum Essential Medium (Mod.) (MEM), Corning
Minimum Essential Medium (MEM), powder with Earle's salts, without L-glutamine, phenol red, and sodium bicarbonate
CAS Number:  
Price
$74.80
Log in for availability
Each (10l)
Minimum order quantity is 1
Product Details & Documents
Minimum Essential Medium (Mod.)(EMEM) based on Basal Medium Eagle (BME) formulation to aid cell propagation.
  • Low endotoxin - < 0.25EU/mL
  • Mycoplasma tested
  • Liquid media products are formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment
It is a common cell culture medium developed from early work using Basal Medium Eagle (BME) with normal mammalian fibroblasts and certain subtypes of HeLa cells. When supplemented with serum, MEM has been used for cultivation of a wide variety of cells grown in monolayers, such as fibroblasts. This formulation contains Earle's salts and with or without L-glutamine.
Specifications
Specifications
  • Description:
    Minimum Essential Medium (MEM), powder with Earle's salts, without L-glutamine, phenol red, and sodium bicarbonate