Order Entry
Canada
Orders LinkContactUsLinkComponent
Corning® Medium 199 (Mod.), Corning

Corning® Medium 199 (Mod.), Corning

Medium 199 (Mod.), Powder [+] Earle's salts [-] sodium bicarbonate, L-glutamine, phenol red, Powder
Catalog # CA45000-574
Supplier:   Corning
Supplier #:  90-050-PB
Corning® Medium 199 (Mod.), Corning
Catalog # CA45000-574
Supplier:  Corning
CAS Number:  
Corning® Medium 199 (Mod.), Corning
Corning® Medium 199 (Mod.), Corning

Medium 199 (Mod.), Powder [+] Earle's salts [-] sodium bicarbonate, L-glutamine, phenol red, Powder

CAS Number:  
Price
$138.00
Log in for availability
Each (10l)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Medium 199 is often used in vaccine production, virology, and for the culture of various non-transformed cells

  • Low endotoxin - < 0.25EU/mL
  • Mycoplasma tested
  • Liquid media products are formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment

It was originally developed as the result of efforts to create a fully defined, animal origin-free medium. Long-term use of this medium requires serum supplementation.

Specifications

Specifications

  • Media Format:
    Powder
  • Description:
    Medium 199 (Mod.), Powder [+] Earle's salts [-] sodium bicarbonate, L-glutamine, phenol red
  • Cat. No.:
    CA45000-574
  • Storage temperature:
    2...8 °C

Specifications