Corning® MEM (Minimum Essential Medium) Alpha Medium, Corning
Minimum Essential Medium (MEM) Alpha Medium with Earle's salts without ribonucleosides, deoxyribonucleosides, and no L-glutamine, Liquid, 500 ml
Supplier: Corning
Supplier #: 15-012-CV
Corning® MEM (Minimum Essential Medium) Alpha Medium, Corning
Catalog # CA45000-302
Supplier: Corning
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Minimum Essential Medium (MEM) is a common cell culture medium developed from early work using Basal Medium Eagle (BME) with normal mammalian fibroblasts and certain subtypes of HeLa cells.
- Low endotoxin - < 0.25EU/mL
- Mycoplasma tested
- Liquid media products are formulated with cell culture grade WFI water
- Manufactured in a cGMP environment
MEM is modified with higher concentrations of amino acids to more closely approximate the protein composition of mammalian cells. When supplemented with serum, MEM has been used for cultivation of a wide variety of cells grown in monolayers, such as fibroblasts.
Certifications: Triple 0.1 µm sterilized.
Documents
Certificates
Search by Lot Number3D Cell Culture Quick Reference Ordering Guide Corning 3D Cell Culture Quick Reference Ordering Guide Corning 3D Cell Culture.pdf Corning® Shopping list for returning to the bench
Product Literature
Cell Culture Media, Molecular Buffers and Reagents
Additional Product Details
Specifications
Specifications
- Media Format:Liquid
- Description:Minimum Essential Medium (MEM) Alpha Medium with Earle's salts without ribonucleosides, deoxyribonucleosides, and no L-glutamine
- Size:500 ml
- Cat. No.:CA45000-302
- Storage temperature:2...8 °C



