Order Entry
Canada
Orders LinkContactUsLinkComponent
 

VWR® Corks

25 mm, 7, 16 mm, 21 mm
  CA23420-210
   Low Manufacturing Impact
VWR® Corks
  CA23420-210
   Low Manufacturing Impact
VWR® Corks
VWR® Corks

25 mm, 7, 16 mm, 21 mm

 
$499.93
   
Pack of 500
 

XXXX Corks function as a closure for bottles, vials, laboratory and other openings.

  • Regular length taper
  • Assorted sizes

The XXXX premium grade corks have fewer lenticels (crevices) than standard grade

Selected grade of fine textured corks. Sizes 22 through 30 have cemented construction.

This means they are better suited for use as laboratory corks and for transporting many kinds of liquids.

 

 
  • Height:
    25 mm
  • Stopper no.:
    7
  • Top diameter:
    21 mm
  • Bottom diameter:
    16 mm
  • Cat. No.:
    CA23420-210
  • Environmentally Preferable:
    Low Manufacturing Impact
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.