Order Entry
Canada
Orders LinkContactUsLinkComponent
BUFFER SOLN TOT CHLORINE 473ML

BUFFER SOLN TOT CHLORINE 473ML

Catalog # CA22635-11
Supplier:   Hach
Supplier #:  2263511
BUFFER SOLN TOT CHLORINE 473ML
Catalog # CA22635-11
Supplier:  Hach
CAS Number:  
BUFFER SOLN TOT CHLORINE 473ML
CAS Number:  
Price
$83.20
Log in for availability
Each (473ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Documents

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.