Order Entry
Canada
Orders LinkContactUsLinkComponent
Olympic™ Stacking Bins, Metro™

Olympic™ Stacking Bins, Metro™

Bentron bin, Black, 41.9W x 37.5L x 17.8H cm (81/4 x 161/2 x 7")
Catalog # CA22221-326
Supplier:   Metropolitan Wire
Supplier #:  SB91516CAS
Olympic™ Stacking Bins, Metro™
Catalog # CA22221-326
Supplier:  Metropolitan Wire
Olympic™ Stacking Bins, Metro™
Olympic™ Stacking Bins, Metro™

Bentron bin, Black, 41.9W x 37.5L x 17.8H cm (81/4 x 161/2 x 7")

Price
$119.79
Log in for availability
Each
Product Details & Documents

Bins are durable, stackable, and feature high impact strength

  • A flat surface on the front of the bin is provided for easy placement of labels and bar codes
  • All bins have an open hopper for easy access to contents
  • Bins feature a lip on the edge of the rear panel for hanging

Choose from Benstat™ or Bentron™ bins:
Benstat is a static-dissipative polypropylene-based material with a surface resistivity of >105ohms/sq. and <1012ohms/sq. All Benstat bins are blue.
Bentron is a polypropylene-based material with a carbon additive to provide an electrically conductive path through the material. Bentron has a surface resistivity of <105ohms/sq. All Bentron bins are black.

Specifications
  • Color:
    Black
  • Description:
    Bentron bin
  • Dimensions:
    41.9W x 37.5L x 17.8H cm (81/4 x 161/2 x 7")
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.