Order Entry
Canada
Orders LinkContactUsLinkComponent
Mops Buffer, 10X, Teknova

Mops Buffer, 10X, Teknova

10X Mops buffer, sterile solution, 100 ml
Catalog # CA200059-678
Mops Buffer, 10X, Teknova
Catalog # CA200059-678
Supplier:  
CAS Number:  
Mops Buffer, 10X, Teknova

10X Mops buffer, sterile solution, 100 ml

CAS Number:  
Discontinued
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

10X MOPS Buffer for EZ Rich defined medium kit

Repeated freeze thaw causes precipitation and a reduction in cell growth. Aliquot contents upon intial thaw

Suitable for molecular biology, protein chemistry, and biochemical applications.

Specifications

Specifications

  • Description:
    10X Mops buffer, sterile solution
  • Size:
    100 ml
  • Cat. No.:
    CA200059-678
  • Storage temperature:
    Frozen
  • Shipping temperature:
    Frozen

Specifications