Order Entry
Canada
Orders LinkContactUsLinkComponent
Molybdate 3 Reagent Solution, Hach

Molybdate 3 Reagent Solution, Hach

Molybdate 3 Reagent Solution, 100 mL Marked Dropping Bottle, Heteropoly blue, 100
Catalog # CA1995-32
Supplier:   Hach
Supplier #:  199532
Molybdate 3 Reagent Solution, Hach
Catalog # CA1995-32
Supplier:  Hach
CAS Number:  
Molybdate 3 Reagent Solution, Hach
Molybdate 3 Reagent Solution, Hach

Molybdate 3 Reagent Solution, 100 mL Marked Dropping Bottle, Heteropoly blue, 100

CAS Number:  
Price
$76.76
Log in for availability
Each (100ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

For low and ultra-low range silica determination by the Heteropoly Blue method

Hach methods 8282 and 8186.

 : Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions.If necessary, we will change your selected shipping method to accomodate these items.

Specifications

Specifications

  • Chemical method:
    Heteropoly blue
  • Description:
    Molybdate 3 Reagent Solution
  • Size:
    100 mL Marked Dropping Bottle
  • No. of tests:
    100
  • Cat. No.:
    CA1995-32
  • Method:
    Heteropoly Blue

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.