Order Entry
Canada
Orders LinkContactUsLinkComponent
AccuGENE™ Buffers | Avantor

AccuGENE™ Buffers

SSC buffer (20X), 1 L, 7.0
Catalog # CA12001-388
Supplier:   Lonza
Fournisseur #:  51205
AccuGENE™ Buffers
Catalog # CA12001-388
Supplier:  Lonza
CAS Number:  
AccuGENE™ Buffers
AccuGENE™ Buffers

SSC buffer (20X), 1 L, 7.0

CAS Number:  
Price
$90.50
Log in pour la disponibilité
Each
Product Details & Documents

AccuGENE® buffers are prepared with high quality reagents and use 18 megohm water

  • Reliable - Manufactured according to strict quality control standards to ensure lot-to-lot consistency
  • High Quality - No detectable DNase, RNase, or protease activities unless otherwise noted
  • Efficient - Ready-made solutions eliminate experiment preparation time
  • Flexible - Customized solutions are available to meet individual needs
  • Formulation - 3.0 M NaCl, 0.3 M sodium citrate, pH 7.0

Store all reagents at 18 to 26 °C (64 to 79 °F).

They are manufactured according to strict quality control standards to ensure lot-to-lot consistency.

Ordering information: For additional sizes, pH, or concentrations, contact your Avantor sales representative.

Documents

Spécifications

Spécifications

  • Description:
    SSC buffer (20X)
  • Size:
    1 L
  • pH:
    7.0
  • Cat. No.:
    CA12001-388
  • Formulation:
    20X SSC with 3.0 M NaCl and 0.3 M Na3C6H5O7

Spécifications

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.