VIAL 90ML PP W/CAP ST CS400
Catalog # CA11006-232
Supplier: Starplex
Frequently Bought Together
NORM-JECT® Luer Lock Sterile Syringes, Air-Tite Products
Supplier: Air-Tite
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Sodium chloride ≥99.0% ACS analytical reagent, VWR Chemicals BDH®
Supplier: VWR International
Peroxide Test, MQuant®, MilliporeSigma
Supplier: MilliporeSigma
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Ethanol denatured, anhydrous
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Corning® McCoy's 5A (Iwakata & Grace Modification), Corning
Supplier: Corning
Calcium Nitrate 4-Hydrate
Supplier: Ward's Science
Potassium iodide 20% (w/v) in aqueous solution stabilized
Supplier: Ricca Chemical
Single-Use Needles, BD Medical
Supplier: BD