Order Entry
Canada
Orders LinkContactUsLinkComponent
Ampicillin solution 100 mg/mL

Ampicillin solution 100 mg/mL

25 ml
Catalog # CA101414-068
Supplier:   Teknova
Supplier #:  A9626
Ampicillin solution 100 mg/mL
Catalog # CA101414-068
Supplier:  Teknova
CAS Number:  69-53-4
Ampicillin solution 100 mg/mL

25 ml

CAS Number:  69-53-4
Price
$228.46
Log in for availability
Each (25ml)
Minimum order quantity is 1
Product Details & Documents

Ampicillin is used as a selectable marker in many commonly used plasmids. A functional Ampicillin gene will confer resistance to cells containing the plasmid.

  • Inhibits cell wall synthesis by inhibiting formation of the peptidoglycan cross-linkages
  • Acts as a competitive inhibitor of transpeptidase

Inhibits bacterial cell wall synthesis by binding to one or more of the penicillin binding proteins (PBPs) that inhibit the final transpeptidation step of peptidoglycan synthesis in bacterial cell walls, thus inhibiting cell wall biosynthesis.

  • CAS Number:69-53-4
  • Formula:C₁₆H₁₉N₃O₄S
  • MDL Number:MFCD00005175
  • Molecular Weight:349.4 g/mol
Specifications

Specifications

  • Size:
    25 ml

Specifications