Order Entry
Canada
Orders LinkContactUsLinkComponent
LB Agar Ampicillin

LB Agar Ampicillin

LB Agar Plates with Ampicillin-100, 100 µl g/ml, 100 mm
Catalog # CA100220-124
Supplier:   Teknova
Supplier #:  L7204
LB Agar Ampicillin
Catalog # CA100220-124
Supplier:  Teknova
CAS Number:  
LB Agar Ampicillin
LB Agar Ampicillin

LB Agar Plates with Ampicillin-100, 100 µl g/ml, 100 mm

CAS Number:  
Price
$1,239.20
Log in for availability
Pack of 20
Minimum order quantity is 8
Product Details & Documents

All plates are tested with at least 4 different recombinant strains of E.

  • Plates shelf-life dated and guaranteed for 4 weeks from date of shipment

  • Must be stored at 4 °C and in absence of continuous exposure to light
  • With proper storage, most antibiotic plates are useable for up to 2 months
  • All plates sold in sleeves of 20 unless noted otherwise

Luria-Bertani plates contain 1% tryptone, 0.5% yeast extract, 1.0% NaCl, and 1.5% agar unless otherwise indicated. LB medium is adjusted to a final pH of 7.0 unless otherwise indicated. All plates are tested with at least 4 different recombinant strains of E. coli to demonstrate the efficacy of the antibiotics and colorimetric indicators contained within.

Coli to demonstrate the efficacy of the antibiotics and colorimetric indicators contained within.

Specifications

Specifications

  • Description:
    LB Agar Plates with Ampicillin-100
  • Fill volume:
    100 µl g/ml
  • Plate size:
    100 mm

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.