Order Entry
Canada
Orders LinkContactUsLinkComponent
 

EMB Agar Plates

EMB (Eosin Blue) Agar, 100 mm, 20 Plates
Catalog # CA100219-774
Supplier:   Teknova
 :  E0070
EMB Agar Plates
Catalog # CA100219-774
Supplier:  Teknova
CAS Number:  
EMB Agar Plates

EMB (Eosin Blue) Agar, 100 mm, 20 Plates

CAS Number:  
Price
$834.82
Log in for availability
Pack of 20
 
Product Details & Documents

EMB (Eosin Blue) agar plates are used as a slightly selective and differential plating medium for the isolation of gram-negative enteric bacteria

  • Sterile
  • Final pH = 7.1 ±10.2 (at 25 °C)

These plates are commonly used as bacterial indicator plates to monitor fermentation. It contains 1.0% Peptone (10.0 g/L), 2.0% Lactose (20.0 g/L), 0.2% Potassium Phosphate, K2HP04 (5.0 g/L), 0.04% Eosin Y (0.4 g/L), 0.0065% Methylene Blue (0.065 g/L), 1.5% Agar (15.0 g/L).

Specifications

Specifications

  • Description:
    EMB (Eosin Blue) Agar
  • Size:
    20 Plates
  • Plate diameter:
    100 mm

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.