EMB Agar Plates
EMB (Eosin Blue) agar, 150 mm, 20 Plates
Catalog # CA100219-680
Jump to:
Discontinued
Product Details & Documents
EMB (Eosin Blue) agar plates are used as a slightly selective and differential plating medium for the isolation of gram-negative enteric bacteria
- Sterile
- Final pH = 7.1 ±10.2 (at 25 °C)
These plates are commonly used as bacterial indicator plates to monitor fermentation. It contains 1.0% Peptone (10.0 g/L), 2.0% Lactose (20.0 g/L), 0.2% Potassium Phosphate, K2HP04 (5.0 g/L), 0.04% Eosin Y (0.4 g/L), 0.0065% Methylene Blue (0.065 g/L), 1.5% Agar (15.0 g/L).
Specifications
Specifications
- Description:EMB (Eosin Blue) agar
- Size:20 Plates
- Plate diameter:150 mm



