Order Entry
Canada
Orders LinkContactUsLinkComponent
CTAB Extraction Solution, Teknova

CTAB Extraction Solution, Teknova

CTAB extraction solution, sterile, 250 ml
Catalog # CA100219-610
CTAB Extraction Solution, Teknova
Catalog # CA100219-610
Supplier:  
CAS Number:  
CTAB Extraction Solution, Teknova

CTAB extraction solution, sterile, 250 ml

CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
BUFFER EXTRCT CTAB 60ML Catalog #CA89167-780

Product CA89167-780 is a replacement for CA100219-610 and has the same packaging and specifications, but may be a different quantity.

BUFFER EXTRCT CTAB 125ML Catalog #CA89167-782

Product CA89167-782 is a replacement for CA100219-610 and has the same packaging and specifications, but may be a different quantity.

Product Details & Documents

CTAB is a cationic detergent used in the extraction of genomic DNA from bacteria and for the isolation of high molecular weight DNA from plants for plamid minipreps and for PCR.

The Solution contains 1% CTAB, 100 mM Tris-HCl (pH 8,0), 20mM EDTA (pH 8,0) and 1.4M Sodium Chloride.

Documents

Specifications

Specifications

  • Description:
    CTAB extraction solution, sterile
  • Size:
    250 ml

Specifications