Order Entry
Canada
Orders LinkContactUsLinkComponent
LB Agar Plates

LB Agar Plates

LB Agar plates with IPTG and X-gal, 100 mm, 20 plates
Catalog # CA100219-336
Supplier:   Teknova
Supplier #:  L1920
LB Agar Plates
Catalog # CA100219-336
Supplier:  Teknova
CAS Number:  
LB Agar Plates

LB Agar plates with IPTG and X-gal, 100 mm, 20 plates

CAS Number:  
Price
$891.40
Log in for availability
Pack of 20
Minimum order quantity is 8
Product Details & Documents

LB is a nutritionally rich growth medium used for the cultivation of recombinant strains of Escherichia coli and enteric bacteria.

  • Shipping conditions: Ambient
  • Storage conditions: Refrigerate

These sterile pre poured plates are enriched with tryptone, yeast extract, sodium chloride, agar, X-gal and IPTG.

Specifications

Specifications

  • Description:
    LB Agar plates with IPTG and X-gal
  • Size:
    20 plates
  • Plate diameter:
    100 mm

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.