Order Entry
Canada
Orders LinkContactUsLinkComponent
YPD Plates

YPD Plates

YPD Agar plates with Sorbitol and 500 µg/ml Zeocin, 100 mm
Catalog # CA100218-980
Supplier:   Teknova
Supplier #:  Y1040
YPD Plates
Catalog # CA100218-980
Supplier:  Teknova
CAS Number:  
YPD Plates
YPD Plates

YPD Agar plates with Sorbitol and 500 µg/ml Zeocin, 100 mm

CAS Number:  
Price
$4,976.50
Log in for availability
Pack of 20
Minimum order quantity is 8
Product Details & Documents

YPD is a nutritionally-rich, undefined, non-selective media for the growth and maintenance of Saccharomyces cerevisiae.

  • 20 Plates
  • Sterile

Ingredients: 1.0% Yeast Extract (10.0 g/L), 2.0% Peptone (20.0 g/L), 2.0% Glucose (20.0g/L), 2.0% Agar (20.0g/L).

Ambient shipping conditions. Store at room temperature or 2–8°C.

Documents

Specifications

Specifications

  • Description:
    YPD Agar plates with Sorbitol and 500 µg/ml Zeocin
  • Size:
    100 mm

Specifications