Orders LinkContactUsLinkComponent
LB Agar Ampicillin
LB Agar Ampicillin
LB Agar Plates with Ampicillin-100, 50 µl g/ml, 100×100 mm
Catalog # CA100218-186
Supplier #:  L3002
LB Agar Ampicillin
Catalog # CA100218-186
Supplier:  
CAS Number:  
LB Agar Ampicillin
LB Agar Ampicillin
LB Agar Plates with Ampicillin-100, 50 µl g/ml, 100×100 mm
CAS Number:  
Discontinued
Product Details & Documents
All plates are tested with at least 4 different recombinant strains of E.
  • Plates shelf-life dated and guaranteed for 4 weeks from date of shipment
  • Must be stored at 4 °C and in absence of continuous exposure to light
  • With proper storage, most antibiotic plates are useable for up to 2 months
  • All plates sold in sleeves of 20 unless noted otherwise
Luria-Bertani plates contain 1% tryptone, 0.5% yeast extract, 1.0% NaCl, and 1.5% agar unless otherwise indicated. LB medium is adjusted to a final pH of 7.0 unless otherwise indicated. All plates are tested with at least 4 different recombinant strains of E. coli to demonstrate the efficacy of the antibiotics and colorimetric indicators contained within.
Coli to demonstrate the efficacy of the antibiotics and colorimetric indicators contained within.
Specifications
Specifications
  • Description:
    LB Agar Plates with Ampicillin-100
  • Fill volume:
    50 µl g/ml
  • Plate size:
    100×100 mm