Order Entry
Canada
Orders LinkContactUsLinkComponent
YPD Plates

YPD Plates

YPD Agar with Sorbitol and 2 mg/mL Zeocin, 100 mm
Catalog # CA100217-508
YPD Plates
Catalog # CA100217-508
Supplier:  
CAS Number:  
YPD Plates
YPD Plates

YPD Agar with Sorbitol and 2 mg/mL Zeocin, 100 mm

CAS Number:  
Discontinued
Product Details & Documents

YPD is a nutritionally-rich, undefined, non-selective media for the growth and maintenance of Saccharomyces cerevisiae.

  • 20 Plates
  • Sterile

Ingredients: 1.0% Yeast Extract (10.0 g/L), 2.0% Peptone (20.0 g/L), 2.0% Glucose (20.0g/L), 2.0% Agar (20.0g/L).

Ambient shipping conditions. Store at room temperature or 2–8°C.

Documents

Specifications

Specifications

  • Description:
    YPD Agar with Sorbitol and 2 mg/mL Zeocin
  • Size:
    100 mm

Specifications

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.