Order Entry
Canada
Orders LinkContactUsLinkComponent
First Strand Buffer, 5X, Teknova

First Strand Buffer, 5X, Teknova

First strand buffer, 5X, 500 ml
Catalog # CA100217-306
Supplier:   Teknova
Supplier #:  F1204
First Strand Buffer, 5X, Teknova
Catalog # CA100217-306
Supplier:  Teknova
CAS Number:  
First Strand Buffer, 5X, Teknova

First strand buffer, 5X, 500 ml

CAS Number:  
Price
811,79 $
Log in for availability
Each (500ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

First strand synthesis buffer designed for use in reverse transcription.

Consists of Tris-HCl (250 mM), pH 8.3, potassium chloride (375 mM), magnesium chloride (15 mM) and DTT (50 mM).

Specifications

Specifications

  • Description:
    First strand buffer, 5X
  • Size:
    500 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.