Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Storage Vials, Quick/Snap, MicroSolv

Vials, quick snap storage, 10.00 ml, 50×22 mm, No, ND18
Catalog # 97058-132
 :  97010-CV10
Storage Vials, Quick/Snap, MicroSolv
Catalog # 97058-132
Storage Vials, Quick/Snap, MicroSolv
Storage Vials, Quick/Snap, MicroSolv

Vials, quick snap storage, 10.00 ml, 50×22 mm, No, ND18

Price
83,18 $
Log in for availability
Pack of 100
 
Product Details & Documents

These easy to use storage vials and caps use a push on technology for ease and comfort.

  • Easy to handle
  • Quick and easy
  • No liners required in the cap

Use these vials to store samples that are not critical.

 : Caps sold separately.

Specifications
  • Cap included:
    No
  • Cap Size:
    ND18
  • Capacity:
    10.00 ml
  • Description:
    Vials, quick snap storage
  • Diameter ×H:
    50×22 mm