Order Entry
Canada
Orders LinkContactUsLinkComponent
Storage Vials, Microsolv™

Storage Vials, Microsolv™

Autosampler Vials, 12.00 ml, 66×18.5 mm, Borosilicate glass, Clear, No, 15-425
Catalog # 97057-156
Supplier #:  97015-CV-12
Storage Vials, Microsolv™
Catalog # 97057-156
Storage Vials, Microsolv™
Storage Vials, Microsolv™

Autosampler Vials, 12.00 ml, 66×18.5 mm, Borosilicate glass, Clear, No, 15-425

Price
$112.85
Log in for availability
Pack of 100
Product Details & Documents

Screw top vials made up of high quality glass for long term storage.

  • Can be used for EPA testing
  • Vials and caps are used to store chemical compounds for years
Specifications
  • Cap included:
    No
  • Cap Size:
    15-425
  • Capacity:
    12.00 ml
  • Color:
    Clear
  • Description:
    Autosampler Vials
  • Material:
    Borosilicate glass
  • Diameter ×H:
    66×18.5 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.