Order Entry
Canada
Orders LinkContactUsLinkComponent
Basik™ 8 mm Screw Top Vials and Inserts, Microsolv

Basik™ 8 mm Screw Top Vials and Inserts, Microsolv

Screw Top Vials, Tear Drop, 1.10 ml, Clear, No, 8-425
Catalog # 97057-138
Supplier #:  9582S-11-0CV
Basik™ 8 mm Screw Top Vials and Inserts, Microsolv
Catalog # 97057-138
Basik™ 8 mm Screw Top Vials and Inserts, Microsolv
Basik™ 8 mm Screw Top Vials and Inserts, Microsolv

Screw Top Vials, Tear Drop, 1.10 ml, Clear, No, 8-425

Price
$210.17
Log in for availability
Pack of 100
Product Details & Documents

These vials are used with Beckman™, CTC™, Gilson™, Knauer™, Shimadzu™, Spark™, Varian™ and Hitachi™ autosamplers that require a narrow neck dimensions.

  • Type-1, N51A Expansion borosilicate glass
  • High quality, low cost with reliable performance

Useful in GC or HPLC.

Specifications
  • Cap included:
    No
  • Cap Size:
    8-425
  • Capacity:
    1.10 ml
  • Color:
    Clear
  • Description:
    Screw Top Vials, Tear Drop
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.