Caps, BASIK™, MicroSolv
PE, White Silicone Rubber / Natural PTFE, 11 mm, Yellow, Snap cap
Catalog # 97055-036
Supplier: MicroSolv Technology Corp
Supplier #: 9502C-E-15Y
Jump to:
Product Details & Documents
Specifications
- Cap Size:11 mm
- Cap type:Snap cap
- Septum:White Silicone Rubber / Natural PTFE
- Material:PE
- Cap color:Yellow