Order Entry
Canada
Orders LinkContactUsLinkComponent
Headspace Caps, MicroSolv

Headspace Caps, MicroSolv

Safety Crimp Cap, 20 mm, 10 mm Hole, Aluminum, Silicone/PTFE White/White 45° Shore A, 3.0 mm, Silver
Catalog # 97054-956
Supplier #:  95025-08-2S
Headspace Caps, MicroSolv
Catalog # 97054-956
Headspace Caps, MicroSolv
Headspace Caps, MicroSolv

Safety Crimp Cap, 20 mm, 10 mm Hole, Aluminum, Silicone/PTFE White/White 45° Shore A, 3.0 mm, Silver

Price
$119.10
Log in for availability
Pack of 100
Minimum order quantity is 1
Product Details & Documents

MicroSolv™ headspace caps are available as magnetic and with 4 different septa types to meet needs.

The screw cap eliminates the need for heavy crimping tools and assures that the vials are sealed perfectly every time. The design saves time and money, and ensures a good head space analysis.

Specifications
  • Septum:
    Silicone/PTFE White/White 45° Shore A, 3.0 mm
  • Color:
    Silver
  • Description:
    Safety Crimp Cap
  • Material:
    Aluminum
  • Size:
    20 mm, 10 mm Hole