Orders LinkContactUsLinkComponent
Headspace Caps, MicroSolv
Headspace Caps, MicroSolv
Magnetic Screw Cap, 18 mm Precision Threads, Closed, Aluminum, Silicone Butyl Red/PTFE Grey, 55° Shore A 1.6 mm Septa, Silver
Catalog # 97054-922
Supplier #:  95020-55S-CD
Headspace Caps, MicroSolv
Catalog # 97054-922
Supplier:  
Headspace Caps, MicroSolv
Headspace Caps, MicroSolv
Magnetic Screw Cap, 18 mm Precision Threads, Closed, Aluminum, Silicone Butyl Red/PTFE Grey, 55° Shore A 1.6 mm Septa, Silver
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
MAGNETIC SCRW TOP HEADSPACE VIAL CAPS Catalog #10827-872
Product 10827-872 is similar to 97054-922 but manufacturer and specifications may vary.
Product Details & Documents
MicroSolv™ headspace caps are available as magnetic and with 4 different septa types to meet needs.
The screw cap eliminates the need for heavy crimping tools and assures that the vials are sealed perfectly every time. The design saves time and money, and ensures a good head space analysis.
Specifications
  • Septum:
    Silicone Butyl Red/PTFE Grey, 55° Shore A 1.6 mm Septa
  • Color:
    Silver
  • Description:
    Magnetic Screw Cap
  • Material:
    Aluminum
  • Size:
    18 mm Precision Threads, Closed