Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Basik™ 8 mm Screw Top Vials and Inserts, Microsolv

Screw Top Vials with Write On Patch, 2.00 ml, Amber, No, 8-425
  97054-614
 :  9582S-WAV
Basik™ 8 mm Screw Top Vials and Inserts, Microsolv
  97054-614
Basik™ 8 mm Screw Top Vials and Inserts, Microsolv
Basik™ 8 mm Screw Top Vials and Inserts, Microsolv

Screw Top Vials with Write On Patch, 2.00 ml, Amber, No, 8-425

 
$75.85
   
Pack of 100
 
 

These vials are used with Beckman™, CTC™, Gilson™, Knauer™, Shimadzu™, Spark™, Varian™ and Hitachi™ autosamplers that require a narrow neck dimensions.

  • Type-1, N51A Expansion borosilicate glass
  • High quality, low cost with reliable performance

Useful in GC or HPLC.

 
  • Cap included:
    No
  • Cap Size:
    8-425
  • Capacity:
    2.00 ml
  • Color:
    Amber
  • Description:
    Screw Top Vials with Write On Patch
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.