Screw Caps, MicroSolv™
PP, Red-Orange Silicone Rubber / TEF, 9-425 mm, Blue, Screw cap, solid
Catalog # 97052-772
Supplier: MicroSolv Technology Corp
Supplier #: 9502S-09-SR-K
Jump to:
Product Details & Documents
Specifications
- Cap Size:9-425 mm
- Cap type:Screw cap, solid
- Septum:Red-Orange Silicone Rubber / TEF
- Material:PP
- Cap color:Blue