Screw Caps, MicroSolv™
PP, Fitted Silicone/PTFE, 9-425 mm, Blue, Screw cap, smooth sided, Robo Style
Catalog # 97052-764
Supplier: MicroSolv Technology Corp
Supplier #: 9502S-05-B-M
Jump to:
Product Details & Documents
These blue screw caps are designed for use with 9-425, 12x32 mm vials.
Documents
Specifications
- Cap Size:9-425 mm
- Cap type:Screw cap, smooth sided, Robo Style
- Septum:Fitted Silicone/PTFE
- Material:PP
- Cap color:Blue